Caspase Fluorometric Substrate Set
Supplier: Enzo Life Sciences
850-229-KI01
89155-982EA 870.33 USD
89155-982
Caspase Fluorometric Substrate Set
Proteins and Peptides
Fluorometric substrates for assaying activities of members of caspase family proteases. All substrates are provided in liquid ready-to-use form.
Ordering information: Contents: Contains 125µl (1mM) each of the following substrates:
- Caspase-1 Substrate, Ac-YVAD-AFC
- Caspase-2 Substrate, Ac-VDVAD-AFC
- Caspase-3 Substrate, Ac-DEVD-AFC
- Caspase-5 Substrate, Ac-WEHD-AFC
- Caspase-6 Substrate, Ac-VEID-AFC
- Caspase-8 Substrate, Ac-IETD-AFC
- Caspase-9 Substrate, Ac-LEHD-AFC