Caspase-3 Colorimetric Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-215-KI01
89155-958EA 437.4 USD
89155-958
Caspase-3 Colorimetric Assay Kit, Enzo Life Sciences
Assays Protein Assays
Based on the spectrophotometric detection of the chromophore pNA (p-nitroaniline) after cleavage from the labelled substrate DEVD-pNA.
Delivery information: Kit includes Cell Lysis Buffer (25 mL), 2X Reaction Buffer (2 mL), 4 mM DEVD-pNA (125 µL), 1 M DTT (100 µL), Dilution Buffer (25 mL).