Human Recombinant CD40L, soluble
Supplier: Enzo Life Sciences
850-064-KI01
89155-938EA 898.71 USD
89155-938
Human Recombinant CD40L, soluble
Proteins and Peptides
Can be used to activate human B cells and dendritic cells. Stimulates growth of human B cells. For stimulation of mouse cells via CD40 use MAb to CD40 (mouse) (FGK45) (Prod. No. ALX-805-046).
Ordering information: Contents: 10µg of CD40L, Soluble (human) (recombinant) (Prod. No. ALX-522-015) and 2x50µg of Enhancer for Ligands (Prod. No. ALX-804-034), which increases the biological activity of rhsCD40L at least 1'000-fold.