Anti-CD20 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
810-209-R100
89155-584EA 311.56 USD
89155-584
Anti-CD20 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit, Isotype: IgG

  • For Immunohistochemistry

Recognizes human CD20.

Type: Primary
Antigen: CD20
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR