Anti-TRP1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
810-022-R100
89155-528EA 678.79 USD
89155-528
Anti-TRP1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit, Isotype: IgG

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human trypsin.

Type: Primary
Antigen: TRP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR