Anti-HLA-E Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
805-709-C100
89155-444EA 330.16 USD
89155-444
Anti-HLA-E Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunoprecipitation

Recognizes human native surface-expressed HLA-E, but not denatured heavy chain of HLA-E.

Type: Primary
Antigen: HLA-E
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR