Anti-CEACAM19 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
805-089-C100
89155-390EA 693.39 USD
89155-390
Anti-CEACAM19 Mouse Monoclonal Antibody
Antibodies
CEA-related cell adhesion molecule 19 (CEACAM19) belongs to the carcinoembryonic antigen (CEA) gene family (1). It encodes a putative glycoprotein which is membrane-bound via a transmembrane domain. Like all members of the CEACAM family, the CEACAM19 protein is a single-pass type I membrane protein and consists of a single extracellular N domain. Two isoforms of CEA-CAM19 exist as a result of alternative splicing events. CEACAM19 expression is ubiquitous with highest expression in prostate, uterus, fetal brain, mammary gland, adrenal gland, skeletal muscle, small intestine, and kidney. Additionally, CEACAM19 is highly expressed in breast and prostate cancer cell lines.

  • For ELISA, Flow Cytometry

Recognizes human CEACAM19. Does not cross-react with human CEACAM1, 3, 4, 5, 6, 7, 8, 20, or 21.

Type: Primary
Antigen: CEACAM19
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR