Anti-TNF-related Apoptosis-inducing Ligand Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-296-C100
89153-996EA 501.86 USD
89153-996
Anti-TNF-related Apoptosis-inducing Ligand Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

Recognizes human TRAIL. Does not cross-react with mouse TRAIL.

Type: Primary
Antigen: TNF-related Apoptosis-inducing Ligand
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR