Anti-CTSL Mouse Monoclonal Antibody [clone: CPLH 33/2]

804-291-C100
89153-988EA 533.18 USD
89153-988
Anti-CTSL Mouse Monoclonal Antibody [clone: CPLH 33/2]
Cathepsin L [EC 3.4.22.15] is a lysosomal endopeptidase belonging to the papain family of the closely related cysteine proteases superfamily. The enzyme is ubiquitously distributed in the lysosomes of all cells and seems to be involved in a variety of disorders in which the pathological destruction of the extracellular matrix is one of the main characteristics.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes the epitope G265YGFEST271 of human procathepsin L. Recognizes mature human single or two chain cathepsin L (heavy chain) and human procathepsin L. Cross-reacts with rat cathepsin L and weakly with mouse procathepsin L. Does not cross-react with mature cathepsins B, D, H, K, S and procathepsins B, D, F, S, W.

Type: Primary
Antigen: CTSL
Clonality: Monoclonal
Clone: CPLH 33/2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR