Anti-TRAF5 Mouse Monoclonal Antibody

804-250-C100
89153-924EA 415.05 USD
89153-924
Anti-TRAF5 Mouse Monoclonal Antibody
Host: Mouse, Isotype: IgG1

  • For Western Blot

Recognizes human TRAF5. Detects a band of ~64kDa by Western blot.

Type: Primary
Antigen: TRAF5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR