Anti-RANKL Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-243-C100
89153-916EA 488.6 USD
89153-916
Anti-RANKL Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1 kappa

  • For Immunohistochemistry, Western Blot

Recognizes human and mouse RANKL. Detects a band of ~35-40kDa by Western blot.

Type: Primary
Antigen: RANKL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR