Anti-FASLG Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-232-C100
89153-892EA 572.22 USD
89153-892
Anti-FASLG Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry

Recognizes human FasL.

Type: Primary
Antigen: FASL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR