Anti-NF-ATC2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-215-C100
89153-870EA 791.62 USD
89153-870
Anti-NF-ATC2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and rat NF-ATc2. Detects bands of ~140kDa (phosphorylated NF-ATc2 in resting immune cells) and ~120kDa (fully-dephosphorylated NF-ATc2 in stimulated cells) by Western blot. Does not cross-react with NF-ATc1.

Type: Primary
Antigen: NF-ATC2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR