Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Apurinic/Apyrimidinic Endonuclease Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-213-R100
Mammalian apurinic/apyrimidinic (AP) endonuclease (APE1; APEX; HAP1; Ref-1) is a multifunctional, bipartite enzyme that belongs to the class II AP endonucleases and that plays an important role in numerous, cellular functions, like repairing abasic sites (loss of purine or pyrimidine base) in DNA, regulating the redox state of other proteins that play roles in oxidative signalling, transcription factor regulation (Fos, Jun, NF-κ B, Myb, HIF-1α, CREB, Pac), cell cycle control (p53) and apoptosis. APE1 initiates the DNA base excision repair (BER) by cleaving the DNA immediately adjacent to the 5’ of an AP site to produce a hydroxyl group at the 3’ terminus of an unmodified nucleotide upstream of the nick and a 5’-deoxyribose phosphate moiety downstream. The product of APE1 is further processed by DNA polymerase β to release 5’-deoxyribose phosphate with its intrinsic lyase activity. The nicked DNA is then sealed by DNA ligase I or DNA ligaseIII/XRCC1 to complete this repair process. In addition to the AP endonuclease activity, APE1 also possesses a 3’-5’ DNA exonuclease activity, a 3’-phosphodiesterase activity, a 3’-phosphatase activity, and a RNase H activity. It has been shown that APE1 is capable of excision L-configuration deoxyribonucleoside analogs from the 3’ termini of DNA. Human APE1 gene is located on chromosome 14q 11.2-12.

For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human APE1. Detects a band of ~37kDa by Western blot.

Type: Primary
Antigen: APURINIC/APYRIMIDINIC ENDONUCLEASE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 804-213-R100 89153-866 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol Apurinic/Apyrimidinic Endonuclease
Antigen Name Apurinic/Apyrimidinic Endonuclease
Reactivity Human
Clonality Monoclonal
Antibody Type Primary
Host Mouse
Isotype IgG2a
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Concentration 1mg/ml
Form Liquid. In PBS containing 0.05% sodium azide.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty