Anti-TNFR1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-200-C100
89153-848EA 419.29 USD
89153-848
Anti-TNFR1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunohistochemistry

Recognizes membrane-bound and soluble human TNF-R1. Does not cross-react with mouse TNF-R1.

Type: Primary
Antigen: TNFR1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR