Anti-CD134L Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-138-C100
89153-732EA 467.18 USD
89153-732
Anti-CD134L Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2c

  • For Immunoprecipitation, Western Blot

Recognizes mouse CD134L. Does not cross-react with human CD134L.

Type: Primary
Antigen: CD134L
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR