Anti-PRF1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-135-C100
89153-726EA 425.3 USD
89153-726
Anti-PRF1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunoprecipitation, Western Blot

Recognizes an epitope in the region aa 402-534 of human and mouse perforin.

Type: Primary
Antigen: PRF1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR