Anti-TOLLIP Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-132-C100
89153-720EA 506.41 USD
89153-720
Anti-TOLLIP Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For Immunoprecipitation, Western Blot

Recognizes human and mouse Tollip.

Type: Primary
Antigen: TOLLIP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR