Anti-PLN Mouse Monoclonal Antibody

804-093-R100
89153-654EA 714.07 USD
89153-654
Anti-PLN Mouse Monoclonal Antibody
Host: Mouse, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, rabbit, chicken, sheep, dog, pig and guinea pig PLB. Recognizes an epitope between aa 9-17 of dog PLB. Detects bands of ~25kDa (non-reducing conditions; representing the pentameric form of PLB) and ~5kDa (reducing conditions; representing the monomeric form of PLB) by Western blot.

Type: Primary
Antigen: PLN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR