Anti-NA+/K+ ATPase alpha1 Mouse Monoclonal Antibody

804-082-R200
89153-634EA 516.45 USD
89153-634
Anti-NA+/K+ ATPase alpha1 Mouse Monoclonal Antibody
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Western Blot

Recognizes the alpha1 subunit of human, rat, sheep, pig and dog Na+/K+ ATPase-alpha. Detects a band of ~110kDa by Western blot. Does not cross-react with Xenopus and chicken Na+/K+ ATPase-alpha1 or with rat Na+/K+ ATPase-alpha2 and -alpha3. Recognizes an epitope between aa 496-506 of sheep kidney Na+/K+ ATPase-alpha.

Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR