Anti-HSP70 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-074-R250
89153-618EA 889.85 USD
89153-618
Anti-HSP70 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot, Gel Shift Assay

Recognizes several members of the HSP70 gene family, including HSP70, HSC70, GRP78, and following heat shock, HSP72 in a broad range of species, including human, mouse, avian, frog, fish, Drosophila, and yeast HSP70. Detects bands from ~70 to ~78kDa by Western blot, representing different members of the HSP70 family. 2-D-gel electrophoresis is required to resolve the heat induced form of these proteins from their constitutively expressed counterparts.

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR