Anti-MAPK Mouse Monoclonal Antibody [clone: 12D4]

Supplier: Enzo Life Sciences
804-061-C100
89153-602EA 723.38 USD
89153-602
Anti-MAPK Mouse Monoclonal Antibody [clone: 12D4]
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes the pThr-Glu-pTyr motif of activated human, mouse, rat and dog ERK1 and ERK2. Requires phosphorylation both at the threonine and the tyrosine site and does not cross-react with the non-phosphorylated form of the protein. Does not cross-react with activated JNK or p38.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: MAPK
Clonality: Monoclonal
Clone: 12D4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR