Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ryanodine Receptor Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-015-C100
The ryanodine receptor (RyR) is the channel responsible for the release of calcium from the sarcoplasmic reticulum (SR) in muscle cells and also plays a role in calcium regulation in non-muscle cells. The RyR exists as a homotetramer and is predicted to have a short cytoplasmic C-terminus and 4-10 transmembrane domains; the remainder of the protein, termed the "foot" region is located in the cytoplasm between the T-tubule and the SR. The mammalian RyR is the product of three different genes: RyR-1, which is expressed predominantly in skeletal muscle and areas of the brain, RyR-2, which is expressed predominantly in the heart muscle but also found in the stomach, endothelial cells and diffuse areas of the brain, and RyR-3 which is found in smooth muscle and the brain (striatum, thalamus and hippocampus).

For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, dog, rabbit, guinea pig, chicken, amphibian and fish RyR-2. Detects a band of 565kDa by Western blot. In non-mammalian vertebrates, a doublet is seen at 565kDa representing the alpha and beta isoforms of the receptor. Cross-reacts weakly with RyR-1.

Type: Primary
Antigen: RYANODINE RECEPTOR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 804-015-C100 89153-534 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol RyRs
Antigen Name Ryanodine Receptor
Reactivity Human
Clonality Monoclonal
Antibody Type Primary
Host Mouse
Isotype IgG1
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
Antigen Synonyms RYR 2, RYDR, Ryanodine receptor 1, Skeletal muscle calcium release channel, ARVD 2, RYR, VTSIP., Cardiac muscle type ryanodine receptor, ARVC 2, RYR1, Central core disease of muscle, RYR 1, hRYR 2, hRYR2, Cardiac muscle ryanodine receptor calcium release channel, Arrhythmogenic right ventricular dysplasia 2, HBRR, RYR 3, MHS, Skeletal muscle ryanodine receptor, RYR2, ARVD2, SKRR, Cardiac muscle ryanodine receptor, Skeletal muscle type ryanodine receptor, Brain ryanodine receptor calcium release channel, Ryanodine receptor 1 skeletal, ARVC2, Ryanodine receptor 2, Ryanodine receptor type1, Ryanodine receptor 2 cardiac, MHS1, RYR3, Sarcoplasmic reticulum calcium release channel, Ryanodine receptor 3, CCO, Brain type ryanodine receptor, Type 1 like ryanodine receptor
Concentration 1mg/ml
Form Liquid. Ascites diluted in PBS containing 0.05% sodium azide.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty