Anti-FASLG Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

804-010F-T100
89153-524EA 472.32 USD
89153-524
Anti-FASLG Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Host: Rat, Isotype: IgG2a

  • For Immunocytochemistry

Recognizes mouse FasL.

Type: Primary
Antigen: FASL
Clonality: Monoclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR