Anti-B4GALT1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-339-C050
89153-512EA 297.15 USD
89153-512
Anti-B4GALT1 Mouse Monoclonal Antibody
β-1,4-Galactosyltransferase 1 (B4GALT1) is one of seven B4GALT genes. Human B4GALT1 encodes a type II membrane protein, which is ubiquitously expressed and involved in glycoconjugate biosynthesis. A soluble form (aa 78 to 398), derived from proteolytic cleavage, is a component of lactose synthase in milk that also contains α-lactalbumin. The amino acid sequence of human B4GALT1 is 88%, 87% and 71% identical to that of rat, mouse/canine and chicken.Defects in B4GALT1 are the cause of multisystem disorders present with a wide variety of clinical features, such as disorders of the nervous system development, psychomotor retardation, dysmorphic features, hypotonia, coagulation disorders, and immunodeficiency.

  • For Immunocytochemistry, Western Blot

Recognizes human B4GALT1. Does not cross-react with mouse B4GALT1.

Type: Primary
Antigen: B4GALT1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR