Anti-FURIN Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-017-R100
89153-392EA 429.45 USD
89153-392
Anti-FURIN Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes an epitope in the cysteine-rich region of human furin convertase. Does not cross-react with mouse furin convertase.

Type: Primary
Antigen: FURIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR