Anti-GCR Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-006-C100
89153-378EA 606.36 USD
89153-378
Anti-GCR Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot, Gel Shift Assay

Recognizes human, mouse, rat, sheep, guinea pig, rabbit and yeast GR. Recognizes full-lengh GR, the 17kDa DNA binding trypsin fragment and the 45kDa steroid- and DNA-binding chymotrypsin fragment. Detects a band of ~97kDa by Western blot. Does not cross-react with primate, avian or amphibian GR.

Reconstitute with distilled water.

Type: Primary
Antigen: GLUCOCORTICOID RECEPTOR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR