Anti-CAMK2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
802-009-R100
89153-348EA 625.14 USD
89153-348
Anti-CAMK2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes recombinant and native mouse and rat phosphorylated alpha-subunit of rat CaM kinase II.

Type: Primary
Antigen: CAMK2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR