Anti-ABCG2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-027-C125 801-027-C250
89153-224EA 604.28 USD
89153-224 89153-226
Anti-ABCG2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry

Recognizes an internal epitope of human BCRP. Does not cross-react with human P-glycoprotein, MRP1, MRP2 or MRP5.

Type: Primary
Antigen: ABCG2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR