Anti-Minor Vault P193 Protein Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-025-C250
89153-218EA 627.13 USD
89153-218
Anti-Minor Vault P193 Protein Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunohistochemistry, Western Blot

Recognizes epitope aa 506-510 (VALGK) of human minor vault p193 protein.

Type: Primary
Antigen: MINOR VAULT P193 PROTEIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR