Anti-MRP1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-012-C250
89153-184EA 1067.82 USD
89153-184
Anti-MRP1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes an internal epitope of human MRP1, overexpressed in various human non-P-glycoprotein MDR tumor cell lines. Does not cross-react with human P-glycoprotein, MDR3 P-glycoprotein or human MRP2 (cMOAT) to MRP5.

Type: Primary
Antigen: MRP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR