Super-iODN (class I/II hybrid) (synthetic) endotoxin-free
746-254-C100
89153-122EA 137.77 USD
89153-122
Super-iODN (class I/II hybrid) (synthetic) endotoxin-free
Super-iODN
Inhibitory iODN oligodeoxynucleotide with phosphorothioate backbone. Particularly potent sequence of an inhibitory ODN for in vivo use in rodents (50-150µg per injection): based on class I (ODN H154) and class II (ODN A151) sequences to combine inhibitory activity on TLRs and STAT signaling.
  • TLR9 inhibitor
For a 100µM stock solution, dissolve the total vial content in 170µl endotoxin-free water (included) or PBS (to order separately). To obtain optimal dissolving we recommend the following procedure:
- Add 50% of the solvent and let dissolve for 10 min.
- A