Aflatoxin G2 ≥98% (by TLC)

Supplier: Enzo Life Sciences
Danger

630-106-M001 630-106-M005
89153-004EA 484.95 USD
89153-004 89153-006
Aflatoxin G2 ≥98% (by TLC)
Aflatoxin G2

Hepatotoxic and hepatocarcinogenic mycotoxin.


Soluble in methylene chloride (5 mg/mL), DMSO or methanol.

Formula: C₁₇H₁₄O₇
Storage Temperature: Freezer
CAS Number: 7241-98-7
EINECS: 230-643-4
UN: 3462
ADR: 6.1,I
https://us.vwr.com/stibo/web/std.lang.all/34/14/45403414.jpg

Order Now

Specification Test Results

Source Isolated from Aspergillus flavus
Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR