Cord factor endotoxin-free
Supplier: Enzo Life Sciences
Synonyms:
Trehalose 6,6’-dimycolate
, TDM
581-210-M001
89152-798EA
319.44
USD
89152-798
Cord factor endotoxin-free
Cord factor
Immunomodulator.
Immunomodulatory compound/adjuvant. Induces activation of macrophages independent of TLR2 or TLR4.
Soluble in chloroform:methanol:water (90:10:1) (5 mg/mL), hexane or isopropanol.
Storage Temperature:
Refrigerator |
CAS Number:
61512-20-7 |
Specification Test Results
Source | Isolated from M. tuberculosis |
Formulation | Lyophilised |
Purity | Single spot (TLC) |
Endotoxin Content |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...