Amiloride, 5-(N,N-Dimethyl)-, hydrochloride ≥97% (by TLC)

550-261-M005
89152-414EA 76.62 USD
89152-414
Amiloride, 5-(N,N-Dimethyl)-, hydrochloride ≥97% (by TLC)
Amiloride, 5-(N,N-Dimethyl)-, hydrochloride

Na+/H+ antiporter inhibitor (IC50=6.9 µM) with increased selectivity and potency relative to amiloride.


  • Na+/H+ antiporter inhibitor


Yellow solid.


Soluble in methanol (10mg/ml) or water (6mg/ml).

Storage Temperature: Refrigerator
CAS Number: 1214-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR