FPL 64176 ≥98%

Synonyms: 2,5-Dimethyl-4-[2-(phenylmethyl)benzoyl]-1H-pyrrole-3-carboxylic acid methyl ester

550-229-M005
89152-366EA 106.57 USD
89152-366
FPL 64176 ≥98%
FPL 64176

Ca²⁺ channel activator.


A potent L-type Ca²⁺ channel opener. Induces a contractile response in cardiac and vascular smooth muscle, EC₅₀=0.2 µM.


Hygroscopic and soluble in 100% ethanol (2 mg/mL) or DMSO (10 mg/mL); insoluble in aqueous solution.


Synonym: 2,5-Dimethyl-4-[2-(phenylmethyl)benzoyl]-1H-pyrrole-3-carboxylic acid methyl ester

Formula: C₂₂H₂₁NO₃
Storage Temperature: Refrigerator
CAS Number: 120934-96-5

Order Now

Specification Test Results

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR