Amiloride hydrochloride dihydrate ≥98%

Supplier: Enzo Life Sciences

550-212-G001
89152-354EA 74.81 USD
89152-354
Amiloride hydrochloride dihydrate ≥98%
Amiloride hydrochloride dihydrate

Ca2+ channel inhibitor


Selectively blocks low threshold (T-type) calcium channel (KD=30μM) with no effect on L-type channels or tetrodotoxin Na+ channels. Inhibits the epithelial tetrodotoxin insensitive Na+ channel (IC50 =1μM). Clinically useful K+ sparing diuretic.

Formula: C₆H₈ClN₇O·HCl·2H₂O
MW: 302.116 g/mol
MDL Number: MFCD00058197
CAS Number: 17440-83-4
https://us.vwr.com/stibo/web/std.lang.all/08/19/45390819.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR