ASSA550-004
550-004-G005
89151-988EA 104.55 USD
89151-988
ASSA550-004
NMDA receptor activator
  • Formulation: White to off-white powder.
Spermidine is a precursor of spermine. It binds to the polyamine modulatory site of the NMDA receptor and has shown to potentiate NMDA-induced currents in a concentration-dependent manner.