Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Omentin (rat), (recombinant), Enzo Life Sciences
Supplier: Enzo Life Sciences
522-132-C010
89151-940EA 460 USD InStock
89151-940
Omentin (rat), (recombinant), Enzo Life Sciences
Proteins and Peptides
Produced in HEK 293 cells. Rat omentin (aa 20-313) is fused at the N-terminus to a FLAG®-tag.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty