ASSA500-002
500-002-L100 500-002-L005 500-002-L025
89151-624EA 81.92 USD
89151-624 89151-626 89151-628
ASSA500-002
Non-ionic detergent.
n-Octyloligooxyethylene is used as a detergent for solubilisation, purification and crystallization of membrane proteins.