Guanosine 5'-O-(2-thiodiphosphate) trilithium salt ≥85% (by HPLC)
480-081-M005
89151-570EA 153.37 USD
89151-570
Guanosine 5'-O-(2-thiodiphosphate) trilithium salt ≥85% (by HPLC)
Guanosine 5'-O-(2-thiodiphosphate) trilithium salt
G protein inhibitor
Cell impermeable, non-hydrolyzable analog of guanosine 5'-O-diphosphate (GDP) that competitively inhibits G-protein activation by GTP and GTP analogs. Use at concentrations similar to GTP-gamma-S (Prod. No. ALX-480-056) (50 µM).
CAS Number: 97952-36-8