Alfacalcidol ≥97%

Supplier: Enzo Life Sciences

Synonyms: 1alpha-Hydroxycholecalciferol1alpha-Hydroxyvitamin D3

460-036-M001
89151-456EA 119.11 USD
89151-456
Alfacalcidol ≥97%
Alfacalcidol

Vitamin D3 precursor


Converts to active calcitriol (1alpha,25-dihydroxyvitamin D3; Prod. No. BML-DM200) in vivo. Inhibits bone resorption.

Formula: C₂₇H₄₄O₂
MW: 400.65 g/mol
MDL Number: MFCD00868328
CAS Number: 41294-56-8
https://us.vwr.com/stibo/web/std.lang.all/07/54/45390754.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR