Pyridoxal-5'-phosphate monohydrate ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydratePyridoxal 5-phosphate monohydratePyridoxal-5-phosphate hydrate

460-011-G005
89151-416EA 271.28 USD
89151-416
Pyridoxal-5'-phosphate monohydrate ≥98% (by HPLC)
Pyridoxal 5'-phosphate monohydrate

  • Formulation: Off-white to beige powder.

Formula: C₈H₁₀NO₆P·1H₂O
MW: 265.17 g/mol (247.15 anhy)
MDL Number: MFCD00149414
CAS Number: 41468-25-1
https://us.vwr.com/stibo/web/std.lang.all/07/50/45390750.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR