5-Aminosalicylic acid ≥98% (by HPLC)

Synonyms: 5-Amino-2-hydroxybenzoic acidMesalamineMesalazinem-Aminosalicylic acid

430-110-G005
89151-124EA 107.77 USD
89151-124
5-Aminosalicylic acid ≥98% (by HPLC)
5-Aminosalicylic acid

NF-kappaB pathway inhibitor


Inhibits TNF-α regulated IκB degradation and NF-κB activation.

Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00007877
CAS Number: 89-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR