R59 022 ≥99%
Supplier: Enzo Life Sciences
430-027-M005
89150-920EA 125.3 USD
89150-920
R59 022 ≥99%
R59 022
Potentiates PKC activity
Potentiates the activity of protein kinase C (PKC) by inhibiting the diacylglycerol metabolism. Inhibits diacylglycerol kinase without affecting phosphodiesterase or phosphatidyl inositol kinase. It inhibits human platelet DAG kinase in intact cells (IC50=3.6μM) and in isolated membranes (IC50=6.7μM).
CAS Number: 93076-89-2
https://us.vwr.com/stibo/web/std.lang.all/06/47/45390647.jpg