(-)-p-Bromotetramisole oxalate ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 6-Bromolevamisole oxalate(-)-p-Bromolevamisole oxalateS(−)-6-(4-Bromophenyl)-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole oxalate

430-025-M005
89150-918EA 93.3 USD
89150-918
(-)-p-Bromotetramisole oxalate ≥98% (by HPLC)
(-)-p-Bromotetramisole oxalate

Potent alkaline phosphatase inhibitor. Mimics the action of orthovanadate in the potentiation of fluorouracil antiproliferative activity. Cell permeable. 


  • Alkaline phosphatase inhibitor


White powder.


Soluble in water, methanol, DMF or DMSO.

Formula: C₁₁H₁₁BrN₂S·C₂H₂O₄
MW: 373,22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070406
CAS Number: 62284-79-1
https://us.vwr.com/stibo/web/std.lang.all/06/46/45390646.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR