SC-9 ≥98% (by TLC)

Supplier: Enzo Life Sciences
Danger

Synonyms: 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulphonamide

420-014-M001 420-014-M005
89150-710EA 92.26 USD
89150-710 89150-708
SC-9 ≥98% (by TLC)
SC-9

PKC activator.


Protein kinase C (PKC) activator in the presence of Ca²⁺.


Soluble in DMSO.


Synonyms: 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide, N-(6-phenylhexyl)-5-chloro-1-naphthalenesulfonamide

Formula: C₂₂H₂₄ClNO₂S
MW: 402.0 g/mol
Storage Temperature: Freezer
CAS Number: 102649-78-5
https://us.vwr.com/stibo/web/std.lang.all/06/14/45390614.jpg

Order Now

Specification Test Results

Purity ≥98% (TLC)
Identity Determined by NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR