4'-Amino-6-hydroxyflavone ≥95% (by TLC)

Synonyms: Aminogenistein

385-021-M001
89150-522EA 323.89 USD
89150-522
4'-Amino-6-hydroxyflavone ≥95% (by TLC)
4'-Amino-6-hydroxyflavone

Inhibitor of the p56lck protein-tyrosine kinase.


  • p56lck tyrosine kinase inhibitor


Yellow powder.


Soluble in DMSO or methanol.

Formula: C₁₅H₁₁NO₃
MW: 253.3 g/mol
Storage Temperature: Freezer
CAS Number: 132018-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR