(+)-Catechin monohydrate ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: (+)-3,3’,4’,5,7-Flavanpentol.H₂O

385-017-G001
89150-516EA 52.02 USD
89150-516
(+)-Catechin monohydrate ≥98% (by HPLC)
(+)-Catechin monohydrate

Free radical scavenger.


Antioxidant flavonoid. Free radical scavenger. Has chemopreventive and antitumor properties.


Soluble in 100% ethanol (50 mg/mL).


Synonym: (+)-3,3’,4’,5,7-Flavanpentol.H₂O

Formula: C₁₅H₁₄O₆·H₂O
MW: 290.3 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00075649
CAS Number: 225937-10-0
EINECS: 205-825-1
https://us.vwr.com/stibo/web/std.lang.all/05/81/45390581.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR