Kaempferol ≥90%

Supplier: Enzo Life Sciences

Synonyms: 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

385-005-M050 385-005-M010
89150-488EA 195.7 USD
89150-488 89150-486
Kaempferol ≥90%
Kaempferol

Fatty acid synthase inhibitor


Antioxidant flavonoid. Apoptosis inducer. Reversible inhibitor of fatty acid synthase (FAS).

Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
MDL Number: MFCD00016938
CAS Number: 520-18-3
https://us.vwr.com/stibo/web/std.lang.all/05/74/45390574.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR